

Lead : Antimony (94:6)%, shot 2 mm (0.08 in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
:
MDL Number:MFCD00134050
Merck Index:12,00733
Boiling Pt:1637 °C (1013 hPa)
Melting Pt:630 °C
Density:6,684 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-100-4
UN:3077
ADR:9,III
:GHS08
:GHS07
Frequently Bought Together