Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Silicon
Silicon ≥99.9% (metals basis), powder APS 1-5 µm
Silicon ≥99.9% (metals basis), powder APS 1-5 µm
:
Thermo Fisher Scientific
Silicon ≥99.9% (metals basis), powder APS 1-5 µm
:
Thermo Fisher Scientific
:
7440-21-3
:
:
Si
:
MFCD00085311
:
28.09 g/mol
:
12,08635
:
Ambient
:
231-130-8
:
1346
:
4.1,III
:
GHS02
ALFA44185.18_TDS_EN
msds_ALFA044185
Thermo Scientific Acroseal 25 Years