Order Entry
Germany
Orders LinkContactUsLinkComponent
Graphite, Sigma-Aldrich®

Graphite, Sigma-Aldrich®

Glass bottle for solids, 113,4 g
Catalog # SIAL496588-113.4G
Supplier:   Merck
Graphite, Sigma-Aldrich®
Catalog # SIAL496588-113.4G
Supplier:  Merck
CAS Number:  7782-42-5
Graphite, Sigma-Aldrich®

Glass bottle for solids, 113,4 g

CAS Number:  7782-42-5
Price
136,00 €
Log in for availability
1 * 113,4 g
Product Details & Documents
  • CAS Number:7782-42-5
  • Formula:C
  • MDL Number:MFCD00144065
  • Molecular Weight:12.01 g/mol
  • Merck Index:12,04560
  • Melting Pt:3827 °C
  • Density:2.09…2.23 g/cm³ (20 °C)
  •  :M-Clarity™ quality level = MQ100
  • EINECS:231-955-3
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    113,4 g

Specifications