Order Entry
Germany
ContactUsLinkComponent
Charcoal activated
Charcoal activated
Catalog # HACH2052626
Supplier:  Hach
CAS Number:  
Charcoal activated
Catalog # HACH2052626
Supplier:  Hach
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :12.01 g/mol
Merck Index:13,01818
Melting Pt:3550 °C
Density:2 g/cm³ (20 °C)
EINECS:231-153-3
UN:1362
ADR:4.2,III
REACH:01-2119488894-16
CAS Number:7440-44-0
 :64365-11-3
Formula:C
MDL Number:MFCD00133992