Charcoal activated -4+8 mesh
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-44-0
:64365-11-3
Formula:C
MDL Number:MFCD00133992
:12.01 g/mol
Merck Index:13,01818
Melting Pt:3550 °C
Density:2 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-153-3
UN:1362
ADR:4.2,III
REACH:01-2119488894-16