Order Entry
Germany
ContactUsLinkComponent
Charcoal activated, NORIT® CN 1 for decolorizing
Charcoal activated, NORIT® CN 1 for decolorizing
Certificates
undefined
Charcoal activated, NORIT® CN 1 for decolorizing
Certificates

Specification Test Results

  • Appearance:
    Black Powder
  • Loss on drying:
    ≤15 %
  • Methylene blue adsorption:
    ≥25.0 g/100 g
  • pH:
    5.5 to 8
  • Phosphate (PO4):
    ≤3.5 % (by acid extraction)
  • Calcium (Ca):
    ≤200 ppm (by acid extraction)
  • Iron (Fe):
    ≤150 ppm (by acid extraction)

Specification Test Results

Related Information