Charcoal activated, DARCO® 4-12 mesh, decolorizing
:7440-44-0
:64365-11-3
:C
:MFCD00133992
:12.01 g/mol
:13,01818
:3652 °C
:2 g/cm³ (20 °C)
:Ambient
:231-153-3
:4.2,III
:01-2119488894-16
- Appearance:Black Granules and/or chunks
- Loss on drying:≤12 %
- Particle size:≤5 % (> 4 mesh)
- pH:3.5 to 6