Charcoal activated, NORIT® SA 2, decolorizing
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-44-0
:64365-11-3
Formula:C
MDL Number:MFCD00133992
:12.01 g/mol
Merck Index:13,01818
Melting Pt:3652 °C
Density:2 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-153-3
ADR:4.2,III
REACH:01-2119488894-16
Specification Test Results
- Appearance:Black Powder
- Loss on drying:≤10 %
- Molasses number:≤390
- Water:≤1.5 %
- GC:≥96.0 %