Aluminium
Catalog # SIALIRMM523B
Supplier: Merck
CAS Number:
Specifications
- Pk:1
- Seq:0001
Specifications
About this item
:
:26.98 g/mol
Merck Index:13,00321
Boiling Pt:2327 °C (1013 hPa)
Melting Pt:660 °C
Density:2.7 g/cm³ (20 °C)
EINECS:231-072-3
CAS Number:7429-90-5
Formula:Al
MDL Number:MFCD00134029