Order Entry
Germany
ContactUsLinkComponent
 
Aluminium
Catalog # SIALIRMM523A
Supplier:  Merck
CAS Number:  
Aluminium
Catalog # SIALIRMM523A
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

 :
 :26.98 g/mol
Merck Index:13,00321
Boiling Pt:2327 °C (1013 hPa)
Melting Pt:660 °C
Density:2.7 g/cm³ (20 °C)
EINECS:231-072-3
CAS Number:7429-90-5
Formula:Al
MDL Number:MFCD00134029