Aluminium, Sigma-Aldrich®
Catalog # SIAL338788-50G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g
- Seq:3368
Specifications
About this item
:
:26.98 g/mol
Merck Index:13,00321
Boiling Pt:2327 °C (1013 hPa)
Melting Pt:660 °C
Density:2.7 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:231-072-3
CAS Number:7429-90-5
Formula:Al
MDL Number:MFCD00134029