Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Aluminium
Aluminium ≥99.999% (metals basis), wire, Puratronic®, annealed, Ø 0.25 mm (0.01 in)
Aluminium ≥99.999% (metals basis), wire, Puratronic®, annealed, Ø 0.25 mm (0.01 in)
:
Thermo Fisher Scientific
Aluminium ≥99.999% (metals basis), wire, Puratronic®, annealed, Ø 0.25 mm (0.01 in)
:
Thermo Fisher Scientific
:
7429-90-5
:
:
Al
:
MFCD00134029
:
26.98 g/mol
:
13,00321
:
2327 °C (1013 hPa)
:
660 °C
:
2.7 g/cm³ (20 °C)
:
Ambient
:
231-072-3
externalurl.pdf
msds_ALFA014445