0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
Aluminium
Aluminium ≥99.9965% (metals basis), rods, Puratronic®, Ø 6 mm (0.2 in)
Aluminium ≥99.9965% (metals basis), rods, Puratronic®, Ø 6 mm (0.2 in)
:
Thermo Fisher Scientific
Aluminium ≥99.9965% (metals basis), rods, Puratronic®, Ø 6 mm (0.2 in)
:
Thermo Fisher Scientific
:
7429-90-5
:
:
Al
:
MFCD00134029
:
26.98 g/mol
:
13,00321
:
2327 °C (1013 hPa)
:
660 °C
:
2.7 g/cm³ (20 °C)
:
Ambient
:
231-072-3
msds_ALFA011478