Aluminium 99.5%, pieces (2-10 mm)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7429-90-5
:
Formula:Al
MDL Number:MFCD00134029
:26.98 g/mol
Merck Index:13,00321
Boiling Pt:2327 °C (760 mmHg)
Melting Pt:660 °C
Density:2.7 g/cm³
Storage Temperature:Ambient
EINECS:231-072-3
Specification Test Results
- Appearance:Grey Pieces
- Particle size:2 to 10 mm
- Total trace metal impurities:≤0.5 %