Aluminium ≥90% stabilised, fine powder, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:7429-90-5
:
Formula:Al
MDL Number:MFCD00134029
:26.98 g/mol
Boiling Pt:2467 °C (1013 hPa)
Melting Pt:660 °C
Density:2.70 g/cm³ (20 °C)
EINECS:231-072-3
UN:1396
ADR:4.3,II
:M-Clarity™ quality level = MQ200
:Stabilised with fat ∼ 2%
:GHS02
Storage Temperature:Ambient
Specification Test Results
- Assay (complexometric):≥ 90.0 %
- Heavy metals (as Pb):≤ 0.03 %
- As (Arsenic):≤ 0.0005 %
- Fe (Iron):≤ 1.0 %
- Fat:passes test