Hydroxyethylcellulose ethoxylate, quaternized, Sigma-Aldrich®
SIAL525944-50G
: Merck
:
- Pack type:Poly bottle
- Pk:50 g
- Seq:3368
- Infrared Spectrum:Conforms to Structure
- Carbon:-%
- Nitrogen:1.5 - 2.2%
- Volatiles:≤6.0%
- Residue on Ignition (Ash):≤3.0%
- Particle Size (-20 Mesh):≥95.0
- Viscosity (C = 2%; Water):300 - 500 cps
:
:290 °C (lit.)
:GHS07
:68610-92-4
:MFCD01771473