ContactUsLinkComponent
Standards for LC performance, test mix, CRM
  REST31699
 :  Restek
 :  
Standards for LC performance, test mix, CRM
  REST31699
 :  Restek
 :  
  • Pack type:
    Ampoule
  • Pk:
    1 Ampoul
  • Description:
    LC performance test mix
  • Pk Info:
    1 ml
Highly effective for characterising LC column parameters and tested against the NIST 870 standard.
  • Mixture of 5 components in methanol
  • Simple, easy, reliable approach to quality control (QC) evaluations or column classification
  • Monitor column performance over time
The test mix contains: amitriptyline hydrochloride (549-18-8) 2,800 µg/ml; ethylbenzene (100-41-4) 1,700 µg/ml; quinizarin (81-64-1) 94 µg/ml; toluene (108-88-3) 1,400 µg/ml; uracil (66-22-8) 28 µg/ml.
 : Meets CRM, ISO requirements.