RIDASCREEN®FAST Enzyme assay, lysozym EIA
Catalog # RBIOR6452
Supplier: R BIOPHARM
CAS Number:
Specifications
- Pk:1 KIT
- Description:RIDASCREEN®FAST Lysozym EIA, 48 wells (6 strips with 8 removable wells each)
Specifications
About this item
RIDASCREEN®FAST Lysozym is a sandwich enzyme immunoassay for the quantitative analysis of lysozyme (hen’s egg protein) in foods like wine, cheese and sausage.