Order Entry
Germany
Orders LinkContactUsLinkComponent
Bridge electrolyte

Bridge electrolyte

Bridge electrolyte, 1 mol/l KNO3, 250 ml bottle, 250 mL
Catalog # METT51350078
Supplier:   Mettler - Toledo
Bridge electrolyte
Catalog # METT51350078
Supplier:  Mettler - Toledo
CAS Number:  
Bridge electrolyte

Bridge electrolyte, 1 mol/l KNO3, 250 ml bottle, 250 mL

CAS Number:  
Price
33,00 €
Log in for availability
1 * 250 mL
Product Details & Documents

Documents

Specifications

Specifications

  • Pk:
    250 mL
  • Description:
    Bridge electrolyte, 1 mol/l KNO3, 250 ml bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.