PromoCell 3D Tumorsphere Medium has been designed to meet your requirements for the extended serial 3D tumorsphere culture. 3D Tumorsphere Medium supports the tumorsphere/mammosphere culture of the most commonly used cancer cell lines.
- Directly switch from 2D to 3D culture
- Supports most commonly used cancer cell lines
- Allows extended serial tumorsphere passages (>10)
- High tumorsphere formation efficiency
- Xeno-free and serum-free defined formulation
- Ready to use
Cell types tested for serial passage with the PromoCell 3D Tumorsphere Medium: Brain U-87 MG, Breast MCF-7; Colon HT-29, Connective tissue HT1080; Liver HepG2; Lung A-549; Pancreas Panc-1; Prostate LNCaP; Skin A-431.
The PromoCell Cancer Stem Cell Medium is ready-to-use and chemically defined providing a culture environment devoid of all stimuli originating from non-defined materials. It is particularly suitable as a cost-efficient tool for the standardized routine-culture of tumorspheres/mammospheres.
List of cell types tested for serial passage with the PromoCell: Bran U-87 MG, Breast MCF-7; Colon HT-29, Connective tissue HT1080; Liver HepG2; Lung A-549; Pancreas Panc-1; Prostate LNCaP; Skin A-431