Osteoblast growth medium
Catalog # C-27015
Supplier: PROMOCELL
CAS Number:
Specifications
- Pk:500 mL
- Description:Osteoblast Basal Medium
Specifications
About this item
PromoCell Osteoblast Growth Medium was developed to establish and maintain human osteoblast cultures. The foetal calf serum in the medium is strictly pre-tested for optimal performance. The media are optimised for primary human cells but can also be used for bovine, murine, and rat osteoblasts as well as osteoblast cell lines.
Ordering information: Ready to use media consists of one bottle of Basal Medium and one vial of SupplementMix. Adding the SupplementMix to the Basal Medium results in the complete Medium. Basal Medium (with or without phenol red) as well as the Osteoblast Growth Medium SupplementMix can also be purchased separately.