Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Cell Culture Supplements and Additives
Supplements for endothelial cell growth medium MV
Supplements for endothelial cell growth medium MV
Supplements for endothelial cell growth medium MV
Catalog # C-39221
Supplier:
PROMOCELL
CAS Number:
Expand image
Supplements for endothelial cell growth medium MV
Catalog # C-39221
Supplier:
PROMOCELL
CAS Number:
Specifications
Pk:
1 SET
Description:
Endothelial Cell Growth Medium MV 2 SupplementPack with separate vials, for 500 ml
Specifications
About this item
Product Details
Product Details