Orders LinkContactUsLinkComponent
Trypsin neutralizing solution
Marketsource
Supplier:   PROMOCELL
Trypsin neutralizing solution
Supplier:  PROMOCELL
Marketsource
Certificates
Trypsin neutralizing solution
Trypsin neutralizing solution
3 Options Available Below
Product Details & Documents
Trypsin is the most widely employed enzyme used for cell detachment. This solution exhibit standardized enzyme activity and contain trypsin from extra-pure lots. This solution contains 0,05% trypsin inhibitor in 0,1% BSA.
 : For in vitro research use only and not for diagnostic or therapeutic procedures.
Product Family Options
Product Information
  • Pk
    Availability
    Price
  • C-41100
    Trypsin neutralising solution (TNS)
    Trypsin neutralising solution (TNS)
    30 mL
    Log in for availability
    Log in for availability
    1 * 30 mL
    1 * 30 mL - 46,00 €
    46,00 €
    1 * 30 mL
       46,00 €
    Specifications:
    Marketsource
    Marketsource
  • C-41110
    Trypsin neutralising solution (TNS)
    Trypsin neutralising solution (TNS)
    125 mL
    Log in for availability
    Log in for availability
    1 * 125 mL
    1 * 125 mL - 114,00 €
    114,00 €
    1 * 125 mL
       114,00 €
    Specifications:
    Marketsource
    Marketsource
  • C-41120
    Trypsin neutralising solution (TNS)
    Trypsin neutralising solution (TNS)
    250 mL
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 209,00 €
    209,00 €
    1 * 250 mL
       209,00 €
    Specifications:
    Marketsource
    Marketsource