Order Entry
Germany
ContactUsLinkComponent
Diprotin A
Diprotin A
Supplier:  ENZO LIFE SCIENCES
Certificates
Diprotin A
Supplier:  ENZO LIFE SCIENCES

Specifications

  • CAS No.:
    90614-48-5
  • MDL No.:
    MFCD00038707
  • Enzyme name:
    Diprotin A
  • Enzyme synonyms:
    Diprotin-A
  • Purity:
    ≥98% (HPLC assay method)
  • Molecular formula:
    C₁₇H₃₁N₃O₄

Specifications