Order Entry
Germany
ContactUsLinkComponent
 
Human recombinant T cell protein tyrosine phosphatase (from E. coli)
  ENZOBMLSE1140200
 :  
Human recombinant T cell protein tyrosine phosphatase (from E. coli)
  ENZOBMLSE1140200
 :  

 

  • Pk:
    1 Vial
  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Seq:
    02
  • Form:
    Liquid
  • Enzyme name:
    T cell protein tyrosine phosphatase
  • Enzyme activity:
    200 U
  • Purity:
    Contains no detectable protease, DNase, RNase, or Ser/Thr phosphatase activity.
  • Formulation:
    In 30% glycerol containing stabilisers.

 

 

Produced in E. coli. T-cell phosphatase with a C-terminal 11 kDa deletion (TCdeltaC11) (aa 1-317).