Human recombinant Matriptase 2
ENZOALX2017521
:
- Pack type:Vial
- Pk:1 Vial
- Enzyme type:Recombinant
- Species:Human
- Seq:02
- Form:Liquid
- Enzyme name:Matriptase 2
- Concentration:100 nM
- Formulation:In 50 mM HEPES, pH 9,0, containing 400 mM sodium chloride and 20% glycerol.
Produced in Drosophila Schneider 2 (S2) cells. Recombinant human matriptase-2 (aa 78-811) is fused at the C-terminus to a V5-His-tag.
- Trypsin-like serine protease.
Matriptase-2 is a potent negative regulator of hepcidin.