Human recombinant Arginase 1
Catalog # ENZOALX201081C020
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:20 µG
- Enzyme type:Recombinant
- Species:Human
- Seq:02
- Enzyme name:Arginase 1
- Purity:≥90% (SDS-PAGE)
- Formulation:In 10 mM TRIS-HCl, pH 7,5, containing 1 mM β-mercaptoethanol, 1 mM MnCl₂ and 50% glycerol.
Specifications
About this item
Produced in E. coli. Full length human arginase I.