Human COP9 signalosome
Catalog # ENZOBMLPW94250020
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:20 µG
- Species:Human
- Seq:02
- Protein synonyms:CSN
- Protein/peptide name:COP9 signalosome
- Formulation:Liquid, in 20 mM TRIS, pH 7,2, containing 10% glycerol, 50 mM KCl and 1 mM mercaptoethanol
Specifications
About this item
Isolated from human erythrocytes.