Order Entry
Germany
ContactUsLinkComponent
Mouse recombinant follistatin
Mouse recombinant follistatin
Catalog # ENZOALX2018140020
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Mouse recombinant follistatin
Catalog # ENZOALX2018140020
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    20 µG
  • Protein/peptide type:
    Recombinant
  • Species:
    Mouse
  • Seq:
    02
  • Biological activity:
    The activity is determined by the dose-dependent neutralization of 7,5 ng/ml human Activin A. Complete neutralization is typically reached at <0,3 µg/ml.
  • Protein synonyms:
    FSH-suppressing protein|FS|Activin-binding protein|FSP
  • Protein/peptide name:
    Follistatin
  • Purity:
    >90% (Reducing and Non-reducing SDS-PAGE)
  • Molecular weight:
    ~31,6kDa
  • Formulation:
    Lyophilized from 10 mM Na2PO4, pH 7,5, containing 50 mM sodium chloride. Sterile filtered.

Specifications

About this item

Produced in E. coli. Non-glycosylated protein, containing 289 amino acids.

  • Carrier-free