Human recombinant Activin-A
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
Produced in E. coli.
- Carrier-free
Non-glycosylated homodimer, containing two 117 amino acid chains.
Specifications
- Protein/peptide type:Recombinant
- Species:Human
- Biological activity:The activity is determined by the ability to induce cytoxicity of MPC-11 cells which is typically observed at concentations <2 ng/ml.
- Protein synonyms:Inhibin beta-1|FRP|FSH-releasing protein
- Protein/peptide name:Activin-A
- Purity:>95% (HPLC, Reducing and Non-reducing SDS-PAGE, UV spectroscopy at 280 nm)
- Molecular weight:~26,2kDa
- Formulation:Lyophilized from a sterile filtered solution containing 0,1% TFA.