Human recombinant SIRT2 (from E. coli), His-tag
Catalog # ENZOBMLSE2510500
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 Vial
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Seq:02
- Tag sequence:HIS Tag
- Protein/peptide name:SIRT2
- Purity:>90% (SDS-PAGE)
- Formulation:Liquid. In 25 mM TRIS, pH 7,5, containing 100 mM NaCl, 5 mM DTT and 10% glycerol.
Specifications
About this item
Produced in E. coli. Contains a N-terminal His-tag.