Order Entry
Germany
ContactUsLinkComponent
CDK5 substrate,Biotin
CDK5 substrate,Biotin
Catalog # ENZOBMLP2640001
Supplier:  Enzo Life Sciences
CAS Number:  
CDK5 substrate,Biotin
Catalog # ENZOBMLP2640001
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Conjugation:
    Biotin
  • Seq:
    02
  • Protein/peptide name:
    CDK5 Substrate
  • Purity:
    >95% (HPLC)
  • Formulation:
    Lyophilized

Specifications

About this item

This peptide, Biotin-Ahx-PKTPKKAKKL, is derived from histone H1 and is a substrate for CDK5.

The biotin allows the peptide to be used in kinase assays with streptavidine-bound membranes.