Histone H3 dimethyl lysine-4 peptide
Catalog # ENZOBMLP2560500
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:0,5 mg
- Seq:02
- Protein/peptide name:Histone H3 dimethyl lysine-4 Peptide
- Purity:95%
Specifications
About this item
Demethylase substrate
Residues 1-21 from the N-terminal tail of human histone H3, dimethylated on the side-chain (epsilon-amino function) of Lys4. Useful as a substrate for histone lysine demethylases, for example LSD1.