Orders LinkContactUsLinkComponent
Casein kinase I substrate peptide
Casein kinase I substrate peptide
Catalog #
Casein kinase I substrate peptide
Catalog #
Supplier:  
CAS Number:  
Casein kinase I substrate peptide
CAS Number:  
Product Details & Documents
This sequence, RRKDLHDDEEDEAMSITA, is derived from inhibitor-2 of protein phosphatase-1 with changes that make it an excellent substrate for casein kinase I (Km=172 µM), while rendering it a poor substrate for casein kinase II.
The addition of two arginyl residues to the amino-terminus allows it to be used in phosphocellulose kinase assays.