Kemptide
Catalog # ENZOBMLP1070001
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 mg
- Seq:02
- Protein/peptide name:Kemptide
- Purity:>97% (HPLC)
- Formulation:Lyophilised
Specifications
About this item
PKA substrate
Phosphate acceptor peptide. Substrate for Ser/Thr protein kinases. Km=4,7 µM for cAMP-dependent protein kinase (PKA). The sequence defines the phosphorylation site of porcine liver pyruvate kinase.