Mouse recombinant LIF (from E. coli)
Catalog # ENZOALX201242L001
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:1 mL
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Mouse
- Seq:02
- Biological activity:Induces macrophage differentiation and proliferation of hematopoietic stem cells; maintains indifferentiated state of of embryonic stem (ES) cell cultures; inhibits proliferation of the mouse myeloid cell line M1.
- Protein/peptide name:LIF
- Purity:>95% (SDS-PAGE)
- Formulation:Liquid. 0,2 μm-filtered solution in PBS containing 0,5% BSA. Contains no preservatives. Further dilutions should be made with medium containing 1% BSA.
- Purification:Purified by FPLC
Specifications
About this item
Produced in E. coli.
Ordering information: Quantity: 107U