Order Entry
Germany
ContactUsLinkComponent
Human recombinant KillerTRAIL™ (soluble) (from E. coli)
Human recombinant KillerTRAIL™ (soluble) (from E. coli)
Catalog # ENZOALX201123C500
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Human recombinant KillerTRAIL™ (soluble) (from E. coli)
Catalog # ENZOALX201123C500
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    500 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Seq:
    02
  • Biological activity:
    Induces apoptosis in a concentration range of 10- 100 ng/ml.Note: This protein does not require a cross-linking enhancer for its potent biological activity. For cell lines that require extensive cross-linking of the TRAIL-Rs for killing (e.g. Jurkat) use Su.
  • Reconstitution instructions:
    For a special storage and dilution buffer see KillerTRAIL™ Storage and dilution buffer (Prod. No. ALX - 505 - 005).
  • Protein/peptide name:
    KillerTRAIL™ (soluble)
  • Purity:
    >98% (SDS-PAGE)
  • Concentration:
    3 mg/ml
  • Formulation:
    Liquid. In 20 mM HEPES, pH 7,4, containing 300 mM sodium chloride, 0,01% Tween 20, 1% sucrose, 1 mM DTT.
  • Purification:
    Affinity purified

Specifications

About this item

Produced in E. coli. The extracellular domain of human TRAIL (aa 95-281) is fused at the N-terminus to a His-tag and a linker peptide.