Dog recombinant Grp94 (from insect cells (Sf9))
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
Produced in Sf21 cells.
Specifications
- Protein/peptide type:Recombinant
- Source:Insect cells (Sf9)
- Species:Dog
- Protein/peptide name:Grp94
- Purity:-
- Formulation:-
- Purification:-