Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
Recombinant GrpE (from
E. coli
)
Recombinant GrpE (from E. coli)
Recombinant GrpE (from E. coli)
Supplier:
ENZO LIFE SCIENCES
Certificates
Expand image
Recombinant GrpE (from E. coli)
Supplier:
ENZO LIFE SCIENCES
About this item
Produced in
E. coli
.
Specifications
Protein/peptide type:
Recombinant
Source:
E. coli
Protein/peptide name:
GrpE
Specifications