Order Entry
Germany
Orders LinkContactUsLinkComponent
Human recombinant HSP70 (from <i>E. coli</i>)

Human recombinant HSP70 (from E. coli)

Supplier:   Enzo Life Sciences
Human recombinant HSP70 (from <i>E. coli</i>)
Supplier:  Enzo Life Sciences
Certificates

2 Options Available Below

Product Details & Documents

Produced in E.coli.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • ENZOADIESP555D
  • ENZOADIESP555F
  • Protein/peptide name:
  • HSP70
  • HSP70
  • Protein/peptide type:
  • Recombinant
  • Recombinant
  • Species:
  • Human
  • Human
  • Source:
  • E. coli
  • E. coli
  • Endotoxin level:
  • Low
  • Low
  • Purity:
  • >90% (SDS-PAGE; Western blot)
  • Formulation:
  • Liquid, In Dulbecco’s PBS containing 2,7 mm potassium chloride, 1,5 mm potassium phosphate, 137 mm sodium chloride and 8,1 mm sodium phosphate.
  • Purification:
  • Purified by multi-step chromatography

Specifications

Product Family Options

Product Information

  • Pk
    Availability
    Price
  • 50 µG
    Log in for availability
    Log in for availability
    1 * 50 µG
    1 * 50 µG - 526,00 €
    526,00 €
    1 * 50 µG
       526,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    ENZOADIESP555D
    Protein/peptide name
    HSP70
    Protein/peptide type
    Recombinant
    Species
    Human
    Source
    E. coli
    Endotoxin level
    Low
    Purity
    >90% (SDS-PAGE; Western blot)
    Formulation
    Liquid, In Dulbecco’s PBS containing 2,7 mm potassium chloride, 1,5 mm potassium phosphate, 137 mm sodium chloride and 8,1 mm sodium phosphate.
    Purification
    Purified by multi-step chromatography
  • 200 µG
    Log in for availability
    Log in for availability
    1 * 200 µG
    1 * 200 µG - 1 320,00 €
    1 320,00 €
    1 * 200 µG
       1 320,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    ENZOADIESP555F
    Protein/peptide name
    HSP70
    Protein/peptide type
    Recombinant
    Species
    Human
    Source
    E. coli
    Endotoxin level
    Low
    Purity
    Formulation
    Purification
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.