Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Protein Linking and Labeling Systems
Red 580 dUTP (lyophilised)
Red 580 dUTP (lyophilised)
Red 580 dUTP (lyophilised)
Catalog # ENZOENZ42844L0050
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Expand image
Red 580 dUTP (lyophilised)
Catalog # ENZOENZ42844L0050
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Specifications
Pk:
50 Nmo
Description:
Red 580 dUTP (lyophilised)
Seq:
02
Purity:
≥93% (HPLC)
Formulation:
Lyophilised
Specifications
About this item
Product Details
Product Details