LSD1 fluorometric drug discovery kit
Catalog # ENZOBMLAK5440001
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 KIT
- Description:LSD1 fluorometric drug discovery kit
- Seq:02
- Size:1 kit
Specifications
About this item
The LSD1 Fluorimetric Drug Discovery Kit provides human recombinant LSD1 and all reagents for measuring its activity in a sensitive, real-time fluorescent assay.
LSD1 catalyzed demethylation of the Histone H3 Dimethyl Lysine-4 Peptide (H3K4Me2 Peptide) produces hydrogen peroxide. A fluorescent signal is generated via the horseradish peroxidase (HRP) catalyzed reaction of the hydrogen peroxide with the CELLestial® Red Peroxidase Substrate. Although fluorescence detection will be more sensitive (Excitation in range of 530-570 nm; Emission ca. 590 nm), the CELLestial® Red peroxidation product may also be detected by absorbance at 563 nm.
Delivery: Recombinant human LSD1 (50µg, >1000U)Histone H3 Dimethyl Lysine-4 Peptide SubstrateCELLestial® Red Peroxidase SubstrateHorseradish PeroxidaseH2O2 StandardTranylcypromine (LSD1 inhibitor)Black and clear ½-vol. 96-well platesDetailed instructions.