CV-Caspase 3 & 7 Detection kit
Catalog # ENZOBMLAK1180001
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:1 KIT
- Description:CV-Caspase 3 & 7 detection kit
- Seq:02
- Storage conditions:4 °C
- Application:Fluorescence Microscopy, Fluorescent Detection
- Size:1 kit
Specifications
About this item
Based on the dye cresyl violet and the caspase 3/7 DEVD peptide recognition sequence, this kit allows the detection of caspases 3 and 7 in living cells by fluorescence microscopy.
Once cells are grown, and apoptosis induced, staining is completed in just a few minutes. Hoechst 33342 and Acridine Orange are also included to allow visualization of nuclei and lysosomes.
A consistant feature of mammalian cell apoptosis is the activation of caspases 3 and/or 7. When cleaved by caspase 3 or 7, the mono and unsubstituted cresyl violet fluoresce when excited at 550-590 nm.
Delivery: Kit contains CR(DEVD)2 reagent, Acridine orange and Hoechst 33342 stain.