Order Entry
Germany
ContactUsLinkComponent
VEGF (Human), ELISA kit
VEGF (Human), ELISA kit
Catalog # ENZOENZKIT1560001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
VEGF (Human), ELISA kit
Catalog # ENZOENZKIT1560001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Conjugate ELISA:
    HRP-Streptavidin
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Human
  • Target protein:
    VEGF
  • Description:
    VEGF (Human), ELISA kit, grow cells
  • Seq:
    01
  • Sensitivity:
    4 pg/ml (range 8 - 2000 pg/ml)

Specifications

About this item

Ultra-sensitive ELISA kit for the quantitative detection of human VEGF with 100% cross reactivity to VEGF 165 and negligible cross-reactivity to VEGF 121.

  • Sensitive Assay: Detect VEGF as low as 4 pg /ml
  • Specific: 100% cross-reactivity to VEGF 165 with negligible cross-reactivity to related molecules including VEGF 121
  • Flexible: Wide dynamic range ideal for various matrices
  • Higher throughput: Assay up to 38 samples in duplicate with each kit with an assay time of just 2.5 hours
  • Efficiency: Color coded reagents help to reduce errors and increase efficiency

For the quantitative determination of human VEGF in serum, plasma, and culture supernatants. The VEGF (human), EIA kit is a sensitive colorimetric immunometric enzyme immunoassay kit with results in just 2,5 hours. Absorbance is read at 450 nm. The wide dynamic range is ideal for various matrices. Assay up to 39 samples in duplicate with each kit.

Ordering information: Contains Microtiter Plate, Conjugate, Detector Antibody, Assay buffer, Antibody Diluent, Wash Buffer Concentrate, Standard, TMB Substrate, Stop Solution.