Order Entry
Germany
ContactUsLinkComponent
HO-1 (rat), ELISA Kit
HO-1 (rat), ELISA Kit
Catalog # ENZOADIEKS810A
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
HO-1 (rat), ELISA Kit
Catalog # ENZOADIEKS810A
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Primary antibody reactivity:
    Rat
  • Target protein:
    HO-1
  • Description:
    HO-1 (rat), ELISA Kit
  • Seq:
    02
  • Sensitivity:
    0,036 ng/ml (range 0,195 - 12,5 ng/ml)

Specifications

About this item

Highly sensitive colorimetric immunometric enzyme immunoassay kit with results in < 3 hours.

For the quantitative determination of rat HO-1 in cell lysates, plasma, serum, and tissue. Cited sample type includes bronchoalveolar lavage fluid.

The HO-1 (rat), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in < 3 hours. Absorbance is read at 450 nm. Save time, money, and precious sample with fully quantitative results and increased sensitivity compared to Western blot analysis.

Ordering information: Contains Microtiter plate, 5X Extraction reagent 2, Standard, Sample diluent, Wash buffer concentrate, Antibody, Conjugate, TMB Substrate, Stop solution 2.