Order Entry
Germany
ContactUsLinkComponent
TGF-β1 ELISA kit
TGF-β1 ELISA kit
Catalog # ENZOADI900155
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
TGF-β1 ELISA kit
Catalog # ENZOADI900155
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Target protein:
    TGF-β1
  • Description:
    TGF-β1 ELISA kit
  • Seq:
    02
  • Sensitivity:
    3,3 pg/ml (range 31,25 - 1000 pg/ml)

Specifications

About this item

For the quantitative determination of TGF-β1 in culture supernatants, plasma, and serum of human, mouse, rat, and cow origin.

The TGF-β1 EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 4 hours. Absorbance is read at 450nm. This kit provides ultra-sensitive measurement of as little as 3,3pg/ml TGF-β1 and has been fully validated in activated serum, plasma, and culture supernatants. The ready-to-use liquid color-coded reagents reduce error.

Ordering information: Contains Microtiter plate, Conjugate, Antibody, Assay buffer 13, Assay buffer 30, Wash buffer concentrate, Standard, TMB Substrate, Stop solution 2.