[pThr183/Tyr185]Jnk1/2 ELISA kit
Catalog # ENZOADI900106
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 KIT
- Target protein:[pThr183/Tyr185]Jnk1/2
- Description:[pThr183/Tyr185]Jnk1/2 ELISA kit
- Seq:02
- Sensitivity:75,8 pg/ml (range 125 - 4,000 pg/ml)
Specifications
About this item
For the quantitative determination of [pThr183/Tyr185]Jnk1/2 in cell lysates of human, mouse, and rat origin.
The [pThr183/Tyr185]Jnk1/2 EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 3 hours. Absorbance is read at 450nm. Save time, money, and precious sample with fully quantitative results and increased sensitivity compared to Western blot analysis.
Ordering information: Contains Microtiter plate, Conjugate, Antibody, Assay buffer 4 concentrate, Wash buffer concentrate, Standard, TMB Substrate, RIPA Cell lysis buffer 2, Stop solution 2.