Order Entry
Germany
ContactUsLinkComponent
[pThr180/Tyr182]p38 ELISA Kit
[pThr180/Tyr182]p38 ELISA Kit
Catalog # ENZOADI900101
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
[pThr180/Tyr182]p38 ELISA Kit
Catalog # ENZOADI900101
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Target protein:
    [pThr180/Tyr182]p38
  • Description:
    [pThr180/Tyr182]p38 ELISA kit
  • Seq:
    02
  • Sensitivity:
    52,1 pg/ml (range 156 - 5,000 pg/ml)

Specifications

About this item

For the quantitative determination of [pThr180/Tyr182]p38 in cell lysates of human and mouse origin.

The [pThr180/Tyr182]p38 EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 3 hours. Absorbance is read at 450nm. Use less precious sample per test compared to Western blot with higher throughput sample analysis; measure up to 40 samples in duplicate per kit.

Ordering information: Contains Microtiter plate, Conjugate, Antibody, Assay buffer 4 concentrate, Wash buffer concentrate, Standard, TMB Substrate, RIPA Cell lysis buffer 2, Stop solution 2.