Order Entry
Germany
ContactUsLinkComponent
 
Protein A ELISA Kit
Catalog # ENZOADI900057
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Protein A ELISA Kit
Catalog # ENZOADI900057
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Description:
    Protein A ELISA kit
  • Seq:
    02
  • Sensitivity:
    9,01 pg/ml (range 15,63 - 1000 pg/ml)

Specifications

About this item

Most sensitive (9,01pg/ml) colorimetric ELISA for monitoring residual contamination.

For the quantitative determination of residual protein A contamination in protein A purified IgG preparations from any species.

The Protein A EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in < 3 hours. Absorbance is read at 450 nm. Ultra-sensitive quantification (< 1 ppm) of Protein A residuals in purified humanized monoclonal antibody preparations. This kit recognizes four different Protein A constructs.

Ordering information: Contains Microtiter plate, Conjugate, Antibody, Assay buffer 13, Wash buffer concentrate,Standard, TMB Substrate, Stop solution 2.