Order Entry
Germany
ContactUsLinkComponent
12(S)-HETE ELISA kit
12(S)-HETE ELISA kit
Catalog # ENZOADI900050
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
12(S)-HETE ELISA kit
Catalog # ENZOADI900050
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Assay Type:
    Competitive
  • Target protein:
    12(S)-HETE
  • Description:
    12(S)-HETE ELISA kit
  • Seq:
    02
  • Sensitivity:
    146 pg/ml (range 195 - 50000 pg/ml)

Specifications

About this item

Sensitive ELISA kit enabling detection of 12(S)-HETE in culture supernatants and plasma from any species.

For the quantitative determination of 12(S)-HETE in culture supernatants and plasma from any species. Cited sample types include serum, renal interstitial fluid and tissue.

The 12(S)-HETE EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 5 hours. Absorbance is read at 405 nm. Fast, accurate, and reliable results are obtained with ready-to-use liquid color-coded reagents that reduce error.

Ordering information: Contains GxR IgG Microtiter plate, Conjugate, Antibody, Assay buffer, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution. Alternative size available: 89141-300 (5x96 wells).